Find methods for your needs
An improved separation of derivatized amino acids using a Thermo Scientific TRACE TR-5 GC column
Description
This method shows the advantages of using the Thermo Scientific TRACE GC Ultra with the Thermo Scientific TRACE TR-5 0.25 µm column for the separation of derivatized amino acids with FID detection.| Market: | Pharma |
| Keywords: | amino acid, Derivatization, L-alanine, L-leucine, L-lysine, MSTFA, N-methyl-N–(trimethylsilyl) trifluoroacetamide, silylation |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.