Find methods for your needs
Application Brief 153: Robust Iso-a-Acids Assaying in Beer within Ten Minutes
Description
The BeerNHop solution for high throughput was applied to analyze beer. Automated analyte enrichment was done by on-line solid phase extraction (SPE) on a Thermo Scientific Hypersil GOLD C8 column. Then, the separation was performed on a Hypersil GOLD 1.9 µm column with UV detection at 270 nm. With a total run time of 9 minutes, the method allows very fast screening for isohumulones in beer. Hence, beer bitterness is quantified and the isohumulone pattern is identified in a matter of minutes. The BeerNHop solution is a valuable tool for the daily business of each brewery.| Market: | Food and Beverage |
| Keywords: | Beer, BeerNHop, Bitterness, Brewery, Brewing, Hop, Isohumulones |
| Matrix: | Beer |
For Research Use Only. Not for use in diagnostic procedures.