Find methods for your needs
Determination of Isohumulones in Beer Be the first to rate this application
Description
Untreated beer was directly injected onto a Thermo Scientific Hypersil GOLD 3 μm column. With a total run time of 15 minutes, the method allows fast screening for isohumulones in beer. Hence, beer bitterness is quantified and the isohumulone pattern is identified within minutes. The BeerNHop solution is a valuable tool for the daily business of each brewery.
Market: Food and Beverage
Keywords: Beer, BeerNHop, Bitterness, Brewery, Brewing, Hop, iso-a-acids, Isohumulones
Matrix: Beer
Uploaded on 10/29/2014.
For Research Use Only. Not for use in diagnostic procedures.