Description
Untreated beer was directly injected onto a Thermo Scientific Hypersil GOLD 3 μm column. With a total run time of 15 minutes, the method allows fast screening for isohumulones in beer. Hence, beer bitterness is quantified and the isohumulone pattern is identified within minutes. The BeerNHop solution is a valuable tool for the daily business of each brewery.| Market: | Food and Beverage |
| Keywords: | Beer, BeerNHop, Bitterness, Brewery, Brewing, Hop, iso-a-acids, Isohumulones |
| Matrix: | Beer |
For Research Use Only. Not for use in diagnostic procedures.