Find methods for your needs
Assay of dexamethasone acetate tablets Assay according to the Chinese Pharmacopoeia using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
In this work the assay of dexamethasone tablets according to the Chinese Pharmacopoeia is performed. The separation is performed using a Thermo Scientific Acclaim 120 C18 HPLC column with UV detection at 240 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Dexamethasone, HPLC, UV detection, Chinese Pharmacopoeia |
| Matrix: | Tablets |
For Research Use Only. Not for use in diagnostic procedures.