Find methods for your needs
The importance of autosampler vial selection in the GC-MS analysis of pyrethroid pesticides at low concentration Be the first to rate this application
importance autosampler vial selection gcms analysis pyrethroid pesticides at low concentration
Description
The selection of the correct autosampler vial type is vital to the success of analyzing pyrethroid pesticides at low levels. A method for the determination of pyrethroids at 0.1 ng/mL was developed using solid phase extraction (SPE) for pre-concentration with subsequent analysis by GC with PTV simulated on-column injection. Careful selection of the autosampler vial type was needed to minimize adsorption effects.
Market: Environmental
Keywords: 33 expansion glass, adsorption, Bifenthrin, Cyfluthrin, Cypermethrin, high purity clear neutral borosilicate glass vial, Permethethrin, Permethrin, Pyrethroid
Matrix: Water
Uploaded on 10/28/2014.
For Research Use Only. Not for use in diagnostic procedures.