Find methods for your needs
Improved separation of Misoprostol and Diclofenac using a Thermo Scientific Hypersil GOLD C18 column
Description
The testing here shows the advantages of using the Thermo Scientific Dionex UltiMate 3000 system; alongside the Thermo Scientific Hypersil GOLD C18 5 µm column for the separation of Misoprostol and Diclofenac with UV detection at 285 nm| Market: | Pharma |
| Keywords: | Diclofenac, HPLC, Hypersil GOLD, Misoprostol, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.