Find methods for your needs
Determination of Tetrahydroisohumulones using Rapid Separation On-Line SPE Be the first to rate this application
Description
The BeerNHop solution for high throughput was applied to analyze the Thermo Scientific Tetrahydroisohumulones standard. Automated analyte enrichment was done by on-line solid phase extraction (SPE) on a Thermo Scientific Hypersil GOLD C8 column. Then, the separation was performed on a Thermo Scientific Hypersil GOLD 1.9 µm column with UV detection at 270 nm. With a total run time of 15 minutes, the method allows fast screening for reduced isohumulones in beer. Hence, beers can be screened if they comply with the German purity law.
Market: Food and Beverage
Matrix: Certified tetrahydroisohumulones standard in methanol (100 µg/mL)
Uploaded on 10/27/2014.
For Research Use Only. Not for use in diagnostic procedures.