Find methods for your needs
USP method for vinorelbine and its impurities using a C18 selectivity HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of vinorelbine and its impurities. The separation was performed on a Thermo Scientific Hypersil GOLD 5µm column with UV detection at 267 nm.| Market: | Pharma |
| Keywords: | Chemotherapy drug, HPLC, Hypersil GOLD, USP, UV detection, Vinorelbine |
| Matrix: | N/A |
| Author: | Anirudha Rakhe |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.