Find methods for your needs
Improved analysis of benzalkonium chloride using a Thermo Scientific Acclaim Surfactant HPLC column Be the first to rate this application
improved analysis benzalkonium chloride using a thermo scientific acclaim surfactant hplc column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of benzalkonium chloride in sterile elastic strip and eye drops. The separation is performed on a Thermo Scientific Acclaim Surfactant HPLC column, with UV detection at 208 nm
Market: Pharma
Keywords: Acclaim Surfactant, Benzalkonium chloride, Eye drops, HPLC, Sterile strips, UltiMate 3000, UV detection
Matrix: Eye drops
Uploaded on 2/22/2010.
For Research Use Only. Not for use in diagnostic procedures.