Find methods for your needs
Amoxicillin capsules assay according to the Chinese Pharmacopoeia using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of amoxicillin capsules according to the Chinese Pharmacopoeia method. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Amoxicillin, Capsule, Chinese pharmacopeia, HPLC, UV detection |
| Matrix: | Capsule |
For Research Use Only. Not for use in diagnostic procedures.