Find methods for your needs
Analysis of paracetamol, caffeine and their impurities using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of paracetamol, caffeine and their impurities. The Thermo Scientific Syncronis C18 5 µm column separated the 5 compounds in less than 13 minutes with resolution between paracetamol and theophylline greater than 3.| Market: | Pharma |
| Keywords: | 4-Aminophenol, 4-Chloroacetanilide, Caffeine, HPLC, Paracetamol, Syncronis C18, Theophylline, UV detection |
| Matrix: | N/A |
| Author: | Anirudha Rakhe |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.