Find methods for your needs
Analysis of benzoic, p-toluic, and terephthalic acids using anion-exchange HPLC
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of residual amounts of benzoic acid and p-toluic acid in excess terephthalic acid. The separation was performed on a Thermo Scientific Hypersil GOLD AX column, with UV detection at 230 nm.| Market: | Food and Beverage |
| Keywords: | Benzoic acid, HPLC, Hypersil GOLD AX, p-toluic acid, terephthalic acid, UltiMate 3000 RSLC, UV detection |
| Matrix: | N/A |
| Author: | Joanne Jones |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.