Description
上述研究结果表明,采用C18 色谱柱分离,柱后加有机相补偿,电雾式检测器检测,获得了较好的分离度与重现性,因此是一个较好的饮料中的三氯蔗糖的分离与检测方法。| Market: | Food and Beverage |
| Keywords: | 三氯蔗糖;液相色谱;电雾式检测器;柱后补偿;饮料 |
| Matrix: | 饮料 |
For Research Use Only. Not for use in diagnostic procedures.