Find methods for your needs
Fast diisocyanates analysis by OSHA Method 42 on a Thermo Scientific Acclaim RSLC PolarAdvantage II HPLC column Be the first to rate this application
fast diisocyanates analysis by osha method 42 on a thermo scientific acclaim rslc polaradvantage ii hplc column
Description
The Thermo Scientific HPLC system is applied for the analysis of diisocyanates by OSHA method 42. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II HPLC column, with UV detection at 254 nm.
Market: Environmental
Keywords: Acclaim PolarAdvantage II (PA2), Diisocyanates, HPLC, OSHA method 42, UV detection
Matrix: N/A
Uploaded on 2/19/2010.
For Research Use Only. Not for use in diagnostic procedures.