Description
本方法在碳酸盐体系淋洗液中加入有机改进剂,使用等度淋洗的方式检测三种甜味剂和两种防腐剂,对仪器配置要求低,分析时间短,灵敏度高,结果准确可靠。适用于白酒、可乐中上述甜味剂和防腐剂的测定。| Market: | Food and Beverage |
| Keywords: | 离子色谱;IonPac AS14;甜味剂;防腐剂;甜蜜素;安赛蜜;糖精钠;山梨酸钾;苯甲酸钠 |
| Matrix: | 食品 |
For Research Use Only. Not for use in diagnostic procedures.