Find methods for your needs
离子色谱-串联质谱法同时测定牛奶中氯酸盐和亚氯酸盐 Be the first to rate this application
离子色谱串联质谱法同时测定牛奶中氯酸盐和亚氯酸盐
Description
本文以IC-MS/MS为检测手段,建立了同时测定亚氯酸盐和氯酸盐的离子色谱方法。方法极大地降低了基体干扰,提高了分析方法的信噪比和灵敏度,该方法应用于牛奶样品中亚氯酸盐和氯酸盐的同时测定,取得了良好的结果。
Market: Food and Beverage
Keywords: 亚氯酸盐;氯酸盐;离子色谱- 串联质谱;牛奶
Matrix: 牛奶
Uploaded on 10/16/2014.
For Research Use Only. Not for use in diagnostic procedures.