Description
婴幼儿食品中的脂肪酸采用乙酰氯甲醇甲酯化法生成脂肪酸甲酯,甲苯提取后进行气相色谱仪测定。该方法操作简单,重复性好,37 种脂肪酸甲酯采用100m 的脂肪酸甲酯分析专用柱,得到了良好的分离。赛默飞世尔科技的Trace1300 系列气相色谱仪用于婴幼儿食品中脂肪酸甲酯的测定灵敏度高,结果准确。| Market: | Food and Beverage |
| Keywords: | 食品;乳品;脂肪酸;气相色谱 |
| Matrix: | 食品 |
For Research Use Only. Not for use in diagnostic procedures.