Description
本方法采用正己烷提取,气相色谱仪检测的技术,建立了多种饮料中16种邻苯二甲酸酯类物质残留的分析方法。该方法具有灵敏、简单快速、准确、线性范围宽等优点,可满足多种类型饮料中邻苯二甲酸酯类物质的检测需要。| Market: | Food and Beverage |
| Keywords: | GC-FID;饮料;邻苯二甲酸酯类物质 |
| Matrix: | 饮料 |
For Research Use Only. Not for use in diagnostic procedures.