Description
本实验采用HPLC-CAD 检测器对庆大霉素有关物质即其C组分进行测定,实验精密度、重复性、线性都较好。与蒸发光测定方法比较,不取对数线性相关系数就可以达0.998,从而计算结果不用进行对数转换,大大简化了含量计算过程。对照品和供试品溶液色谱图中C2 和C2a 之间的峰为小诺霉素峰,它们三个之间的分离度大于2.0,由于该化合物为庆大霉素C 组分中的次要成分所以未对其进行分析。| Market: | Pharma |
| Keywords: | HPLC-CAD;庆大霉素;庆大霉素C 组分 |
| Matrix: | 庆大霉素注射液 |
For Research Use Only. Not for use in diagnostic procedures.