Description
本试验优化了中国药典测定银杏制剂中银杏内酯含量色谱条件,并采用电喷雾检测器(CAD)测定了其含量,可获得比ELSD检测器更好的灵敏度、重现性,以及降低银杏内酯对照品的需求量,大大降低银杏制剂中测定色谱条件的复杂性,提高了方法的适用性,降低了检测成本。| Market: | Pharma |
| Keywords: | 液相色谱;电喷雾检测器;银杏;银杏内酯 |
| Matrix: | 银杏制剂 |
For Research Use Only. Not for use in diagnostic procedures.