Find methods for your needs
Separation of ceftriaxone and sulbactam sodium salts using a simple isocratic reversed-phase method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied with the Thermo Scientific Hypersil GOLD 5 µm column for the separation of ceftriaxone and sulbactam sodium salts in less than 15 minutes with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Antibiotics, Ceftriaxone, HPLC, Hypersil GOLD, Sulbactam, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.