Description
盐霉素在CAD检测器上有较好的响应,进样精密度、重复性、加样回收率都较好。方法准确简便,可用于主成分和杂质的含量测定。根据CAD检测器原理,各种物质在CAD上的响应因子相似,在杂质标准品不能获得的情况下,通过面积归一化法,更能反映各杂质成分和主成分的真实含量。| Market: | Pharma |
| Keywords: | 电喷雾检测器;预混剂;盐霉素;杂质 |
| Matrix: | 兽药 |
For Research Use Only. Not for use in diagnostic procedures.