Description
三种胆碱类化合物在本实验条件下,可以被很好的分离和测定。此方法不需衍生,操作简单,且方法有很好的特异性和重现性,结果可靠,可用于卡巴胆碱注射液含量与有关物质的同时检测。| Market: | Pharma |
| Keywords: | 离子色谱;电导;IonPac CS17;卡巴胆碱注射液;含量测定;有关物质 |
| Matrix: | 注射液 |
For Research Use Only. Not for use in diagnostic procedures.