Find methods for your needs
伊班膦酸钠注射液中磷酸盐与亚磷酸盐测定 Be the first to rate this application
伊班膦酸钠注射液中磷酸盐与亚磷酸盐测定
Description
本方法可同时检测伊班膦酸注射液中的磷酸盐和亚磷酸盐,操作简单,方法重现性较好,结果可靠。
Market: Pharma
Keywords: 离子色谱;电导;IonPac AS18;伊班膦酸钠注射液;有关物质
Matrix: 注射液
Uploaded on 10/15/2014.
For Research Use Only. Not for use in diagnostic procedures.