Find methods for your needs
Improved resolution of anthocyanins and anthocyanidins from bilberry using a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of anthocyanins and anthocyanidins from bilberry. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 520 nm.
Market: Pharma
Keywords: Acclaim 120 C18, Anthocyanidins, Anthocyanins, Cyanidin, Delphinidin, HPLC, Malvidin, Pelargonidin, Peonidin, Petunidin, UV detection
Matrix: Strong acidic bilberry extract
Uploaded on 2/18/2010.
For Research Use Only. Not for use in diagnostic procedures.