Description
采用国标方法、气相色谱质谱联用仪检测的技术,建立了测定24种芳香胺的GC-MS方法。方法的灵敏度高,24种芳香胺检出限分别为0.01,0.02,01,0.2 μg/mL,在1.0、5.0 mg/L两个添加水平下,回收率为78~113%。该方法对偶氮测定具有灵敏、准确、线性范围宽等优点,可满足纺织品中芳香胺类物质的检测需要。| Market: | Chemical |
| Keywords: | 纺织品;芳香胺;气相色谱质谱 |
| Matrix: | 纺织品 |
For Research Use Only. Not for use in diagnostic procedures.