Find methods for your needs
Fast determination of bimatoprost using a simple reversed-phase HPLC-UV method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the analysis of the assay of the active pharmaceutical ingredient Bimatorost. The separation was performed with a Thermo Scientific Accucore aQ 2.6 µm column with UV detection at 210 nm and a total run time of 6 minutes.| Market: | Pharma |
| Keywords: | Accucore aQ, Bimatoprost, HPLC, Prostaglandin, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.