Description
采用赛默飞世尔全新一代TRACE 1310 GC 含氢火焰检测器,结合Triplus-300 顶空自动进样器。Triplus-300 顶空自动进样器具有高达300℃的孵化温度,同时具有120 位的进样盘和18 位的同时孵化炉,可以满足实验室的高样品量及缩短分析时间,提高分析效率的作用。另外其具有安装快捷方便,同时测定灵敏度高、重复性好、结果可靠等优点,本文完全满足饮用水中挥发性有机物的分析与检测需要,同时可以轻松应对实验室各种对水质分析的要求。| Market: | Environmental |
| Keywords: | 顶空法;气相色谱;TG-Wax MS;环境水;挥发有机物 |
| Matrix: | 环境水 |
For Research Use Only. Not for use in diagnostic procedures.