Find methods for your needs
Fast separation of urea herbicides using a reversed-phase HPLC-UV method
Description
The Thermo Scientific HPLC system is applied with the Thermo Scientific Accucore 2.6 µm RP-MS column for the fast analysis of a range of urea herbicides. The separation was achieved in less than four minutes with UV detection showing this column can provide good baseline resolution of the analytes with low operating pressures.| Market: | Environmental |
| Keywords: | Accucore RP-MS, Chlortoluron, Diuron, Linuron, Metoxuron, Monuron, Terbuthiuron, Urea herbicides, UV detection |
| Matrix: | N/A |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.