Find methods for your needs
An improved analysis of glucocorticosteroids using a Thermo Scientific Acclaim Phenyl-1 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of glucocorticosteroids. The Thermo Scientific Acclaim Phenyl-1 3 µm column separated eight glucocorticosteroids in less than 20 minutes with UV detection.| Market: | Pharma |
| Keywords: | 6-Methylprednisolone, Acclaim Phenyl-1, Corticosterone, Cortisone, Deoxyhydrocortisone, Dexamethasone, Glucocorticosteroid, HPLC, Hydrocortisone, Prednisolone, Prednisone, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.