Find methods for your needs
An improved separation of four aflatoxin standards on a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
an improved separation four aflatoxin standards on a thermo scientific acclaim 120 c18 hplc column
Description
The separation of four aflatoxin standards was performed on a Thermo Scientific Acclaim 120 C18 3 µm column in less than 10 minutes with a UV detector.
Market: Environmental
Keywords: Acclaim 120 C18, Aflatoxin, B1, B2, G1, G2, HPLC, UV detection
Matrix: N/A
Uploaded on 7/21/2014.
For Research Use Only. Not for use in diagnostic procedures.