Find methods for your needs
The effect of pH on selectivity on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column Be the first to rate this application
effect ph on selectivity on a thermo scientific acclaim mixedmode wax1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used to demonstrate the pH effect on selectivity. Changing the pH affects retention of weak acids without affecting neutral compounds.
Market: Other
Keywords: 4-Hydroxybenzoic acid, Acclaim Mixed-Mode WAX-1, Butylbenzene, HPLC, pH, Selectivity, UV detection
Matrix: N/A
Uploaded on 7/17/2014.
For Research Use Only. Not for use in diagnostic procedures.