Find methods for your needs
The effect of organic solvent on selectivity on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column Be the first to rate this application
effect organic solvent on selectivity on a thermo scientific acclaim mixedmode wax1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used to demonstrate the organic solvent effect on selectivity. Changing the organic solvent content affects retention of neutral molecules more strongly and in the opposite direction of anions.
Market: Other
Keywords: 4-Hydroxybenzoic acid, Acclaim Mixed-Mode WAX-1, Butylbenzene, HPLC, Organic solvent, Selectivity, UV detection
Matrix: N/A
Uploaded on 7/17/2014.
For Research Use Only. Not for use in diagnostic procedures.