Find methods for your needs
The effect of organic solvent on selectivity on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used to demonstrate the organic solvent effect on selectivity. Changing the organic solvent content affects retention of neutral molecules more strongly and in the opposite direction of anions.| Market: | Other |
| Keywords: | 4-Hydroxybenzoic acid, Acclaim Mixed-Mode WAX-1, Butylbenzene, HPLC, Organic solvent, Selectivity, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.