Find methods for your needs
Acceleration of EPA Method 8330 with a Thermo Scientific Acclaim Explosives E2 column
Description
The separation of 14 explosives using a Thermo Scientific Acclaim Explosives E2 column was performed in less than 12 minutes. This is a threefold acceleration of EPA Method 8330.| Market: | Environmental |
| Keywords: | 1,3,5-TNB, 1,3-DNB, 2,4,6-TNT, 2,4-DNT, 2,6-DNT, 2-NT, 3-NT, 4-Am-2,6-DNT,2-Am-4,6-DNT, 4-NT, Acclaim Explosives E2, EPA Method 8330, HMX, HPLC, NB, RDX, Tetryl |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.