Find methods for your needs
Improved analysis of benzalkonium salts in spray disinfectant using a Thermo Scientific Acclaim Surfactant column
Description
The Thermo Scientific Acclaim Surfactant column is used for the determination of benzalkonium salts in spray disinfectant. Suppressed conductivity detection is adopted, which is specific for cations and does not suffer interference from other ingredients that may be present.| Market: | Pharma |
| Keywords: | Acclaim Surfactant, Benzalkonium salts, Disinfectant spray, Germicide, HPLC, Suppressed conductivity |
| Matrix: | Disinfectant spray |
For Research Use Only. Not for use in diagnostic procedures.