Find methods for your needs
Rapid determination of active ingredients in sunscreen lotion by normal-Phase chromatography using a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
The Thermo Scientific HPLC system is applied to determine active ingredients in sunscreen lotion, which has been prepared for analysis by liquid-liquid extraction. The separation was performed on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column in normal-phase mode with UV detection at 305nm.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, HPLC, Liquid-liquid extraction, Normal-phase, Octyl methoxycinnamate, Octyl salicylate, Sunscreen, UV detection |
| Matrix: | Sunscreen |
For Research Use Only. Not for use in diagnostic procedures.