Find methods for your needs
Fast separation of Nitrophenols using a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column Be the first to rate this application
Description
Nitrophenol compounds are known for their variable retention and poor peak shape when separated using conventional RP-HPLC. The Thermo Scientific Acclaim Mixed-Mode WAX-1 column gives reliable, sharp peaks using UV detection at 360 nm.
Market: Environmental
Keywords: Acclaim Mixed-Mode WAX-1, HPLC, Nitrophenols, UV detection
Matrix: N/A
Uploaded on 5/28/2014.
For Research Use Only. Not for use in diagnostic procedures.