Find methods for your needs
Fast separation of Nitrophenols using a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
Nitrophenol compounds are known for their variable retention and poor peak shape when separated using conventional RP-HPLC. The Thermo Scientific Acclaim Mixed-Mode WAX-1 column gives reliable, sharp peaks using UV detection at 360 nm.| Market: | Environmental |
| Keywords: | Acclaim Mixed-Mode WAX-1, HPLC, Nitrophenols, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.