Find methods for your needs
Fast analysis of glucocorticosteroids using the Thermo Scientific Acclaim C30 HPLC column Be the first to rate this application
Description
Separation of glucocorticosteroids is challenging due to their structural similarity. The Thermo Scientific Acclaim C30 offers improved resolution and throughput with the same elution order under the same conditions compared to a C18 column.
Market: Pharma
Keywords: 11-Deoxyhydrocortisone, 6-Methylprednisolone, Acclaim C30, Corticosterone, Cortisone, Dexamethasone, Glucocorticosteroid, HPLC, Hydrocortisone, Prednisolone, Prednisone, UV detection
Matrix: N/A
Uploaded on 5/28/2014.
For Research Use Only. Not for use in diagnostic procedures.