Find methods for your needs
Accurate profiling of carotenoids in tomato using a Thermo Scientific Acclaim C30 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the analysis of carotenoids in red tomato. The separation is performed on a Thermo Scientific Acclaim C30 3 µm HPLC column, with UV detection at 450 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, beta-carotene, carotenoid, HPLC, Lutein, Lycopene, Tomato, UV detection |
| Matrix: | Red tomato |
For Research Use Only. Not for use in diagnostic procedures.