Find methods for your needs
Fast analysis of terephthalic, benzoic and p-toluic acids using a Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific UltiMate 3000RS HPLC system is applied for the analysis of terephthalic, benzoic and p-toluic acids. The separation is performed on a Thermo Scientific Acclaim Trinity P1 HPLC column, with UV detection at 254 nm.| Market: | Chemical |
| Keywords: | Acclaim Trinity P1, Benzoic acid, HPLC, p-toluic acid, terephthalic acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.