Find methods for your needs
Improved carbonyl-DNPH standards analysis using a Thermo Scientific Acclaim Carbonyl RSLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of carbonyl-DNPH standards. The separation is performed on a Thermo Scientific Acclaim Carbonyl RSLC column, with UV detection at 360 nm.
Market: Environmental
Keywords: Acclaim Carbonyl, Carbonyl-DNPH standards, EPA 554, HPLC, UV detection
Matrix: N/A
Uploaded on 5/8/2014.
For Research Use Only. Not for use in diagnostic procedures.