Find methods for your needs
Improved analysis of 18 polycyclic aromatic hydrocarbons (PAHs) using a Thermo Scientific Hypersil Green PAH HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of polycyclic aromatic hydrocarbons (PAHs). The separation is performed on a Thermo Scientific Hypersil Green PAH HPLC column, with UV detection at 225 nm.| Market: | Environmental |
| Keywords: | HPLC, Hypersil Green PAH, PAHs, Polycyclic aromatic hydrocarbons, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.