Find methods for your needs
Rapid analysis of testosterones using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of testosterones. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Epitestosterone, HPLC, Ketotestosterone, Nortestosterone, Testosterones, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.