Find methods for your needs
Rapid analysis of cetirizine hydrochloride using a Thermo Scientific Accucore HILIC HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of cetirizine hydrochloride. The separation is performed on a Thermo Scientific Accucore HILIC HPLC column, with UV detection at 230 nm.
Market: Pharma
Keywords: Accucore HILIC, Cetirizine hydrochloride, HPLC, UV detection
Matrix: N/A
Uploaded on 2/25/2013.
For Research Use Only. Not for use in diagnostic procedures.