Find methods for your needs
Separation of curcuminoids from turmeric - comparison of polar embedded and C18 solid core HPLC columns Be the first to rate this application
Description
The Thermo Scientific Accucore Polar Premium HPLC column has a polar-embedded C18 surface on a superficially porous silica substrate. This chemistry provides a selectivity that resolves the major and minor components of turmeric under simple isocratic conditions in about three minutes, while C18 columns fail to separate these components.
Market: Food and Beverage
Keywords: Accucore Polar Premium, Bis-desmethoxycurcumin, Curcumin, Curcuminoids, Desmethoxycurcumin, HPLC, UV detection
Matrix: Turmeric root
Uploaded on 2/7/2013.
For Research Use Only. Not for use in diagnostic procedures.