Find methods for your needs
Rapid separation of analgesic compounds using a Thermo Scientific Accucore Urea-HILIC HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of analgesic compounds aspirin, acetaminophen and salicylic acid. The separation is performed on a Thermo Scientific Accucore Urea-HILIC HPLC column, with UV detection at 230 nm.| Market: | Pharma |
| Keywords: | Accucore Urea-HILIC, Acetaminophen, Analgesics, Aspirin, HPLC, Salicylic acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.