Find methods for your needs
Fast analysis of estrogens using a Thermo Scientific Accucore Phenyl-X HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of estrogens. The separation is performed on a Thermo Scientific Accucore Phenyl-X HPLC column, with UV detection at 220 nm.| Market: | Pharma | 
| Keywords: | Accucore Phenyl-X, Estradiol, Estriol, Estrogens, Estrone, Ethynylestradiol, HPLC, UV detection | 
| Matrix: | N/A | 
For Research Use Only. Not for use in diagnostic procedures.